EMAIL THIS PAGE TO A FRIEND
Journal of acquired immune deficiency syndromes (1999)
Impact of hepatitis C virus on HIV response to antiretroviral therapy in Nigeria.
PMID 23196830
Abstract
The effect of hepatitis C virus (HCV) on antiretroviral therapy (ART) response in patients in sub-Saharan Africa is unknown. We studied 1431 HIV-infected ART initiators in Jos, Nigeria, of whom 6% were HCV coinfected. A similar proportion of HIV/HCV-coinfected and HIV-monoinfected patients achieved HIV RNA <400 copies per milliliter after 24 and 48 weeks of ART (P > 0.05). Hepatotoxicity was uncommon (0.8% and 0.33% at 24 and 48 weeks, respectively) but was more common in the HIV/HCV-coinfected group at 24 (adjusted odds ratio = 19.3; 95% confidence interval: 4.41 to 84.4) and 48 weeks (adjusted odds ratio = 56.7; 95% confidence interval: 5.03 to 636.92). HCV did not significantly impact ART response in this Nigerian cohort.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1413
2′,3′-Didehydro-3′-deoxythymidine, ≥98% (TLC)
C10H12N2O4