EMAIL THIS PAGE TO A FRIEND
Blood
Gene therapy. Factor IX Padua: them that have, give.
PMID 23197580
Abstract
In this issue of Blood, Finn et al have taken a factor IX variant with increased specific activity associated with thrombophilia and used it to improve gene therapy of hemophilia B in dogs, and Cantore et al have shown similar results in mice.