EMAIL THIS PAGE TO A FRIEND
Tropical medicine & international health : TM & IH
Increased prevalence of dhfr and dhps mutants at delivery in Malawian pregnant women receiving intermittent preventive treatment for malaria.
PMID 23198734
Abstract
In the context of an Intermittent preventive treatment (IPTp) trial for pregnant women in Malawi, Plasmodium falciparum samples from 85 women at enrollment and 35 women at delivery were genotyped for mutations associated with sulfadoxine-pyrimethamine resistance. The prevalence of the highly resistant haplotype with mutations at codons 51 and 108 of dihydrofolate reductase (dhfr) and codons 437 and 540 of dihydropteroate synthase (dhps) increased from 81% at enrollment to 100% at delivery (P = 0.01). Pregnant women who were smear-positive at enrollment were more likely to have P. falciparum parasitemia at delivery. These results lend support to concerns that IPTp use may lead to increased drug resistance in pregnant women during pregnancy and emphasise the importance of screening pregnant women for malaria parasites in areas with prevalent SP resistance even when they are already on IPTp.