EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of clinical and laboratory investigation
Relationship of BsmI polymorphism of vitamin D receptor gene with left ventricular hypertrophy and atherosclerosis in hemodialysis patients.
PMID 23198772
Abstract
Left ventricular hypertrophy (LVH) is a common manifestation of cardiovascular disease and has an important prognostic value in patients with end-stage renal disease (ESRD). Vitamin D receptor (VDR) has been intensively investigated, and one of these (BsmI) already has been associated with survival in the dialysis population. The aim of this study was to investigate the role of VDR polymorphism (BsmI) on the development of ventricular hypertrophy and atherosclerosis in hemodialysis patients. Subject and methods. The subjects were 80 patients with end-stage renal disease on maintenance hemodialysis, and 40 healthy controls. Clinical and laboratory parameters, including genetic variation in VDR gene (BsmI), were assessed. In addition, echocardiography and intima-media thickness were performed for all subjects. There was no significant difference in the distribution of BsmI genotypes either in patients or in the control group. The frequency of the B allele of BsmI polymorphism (41.6%) in dialysis patients was similar to that of healthy control subjects (39.2%). Patients with BB genotype had significantly lower serum concentrations of 25-hydroxy vitamin D compared to both Bb and bb genotypes. The number of B alleles was positively correlated with left ventricular mass index (LVMI), but not with intima-media thickness. These results suggest that the B alleles of the BsmI polymorphism could be considered as novel markers of altered vitamin D signaling in ESRD patients, and this alteration in BB genotype produces an increase in left ventricle mass.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R6142 Acc I from Acinetobacter calcoaceticus, Restriction Enzyme
R6885 Alu I from Arthrobacter luteus, Restriction Enzyme
R8631
Bcl I from Bacillus caldolyticus, Restriction Enzyme
R6377
Bgl II from Bacillus licheniformis, Restriction Enzyme
R3131
Bln I from Brevibacterium linens, buffered aqueous glycerol solution
R3635
Bsm I from Bacillus stearothermophilus NUB 36, Restriction Enzyme
R4253 BstE II from Bacillus stearothermophilus ET, Restriction Enzyme
R1761
Cfo I from Clostridium formicoaceticum, Restriction Enzyme
R7763
Cla I from Caryophanon latum L, Restriction Enzyme
R4256
Dde I from Desulfovibrio desulfuricans strain Norway, Restriction Enzyme
R4381
Dra I from Deinococcus radiophilus, Restriction Enzyme
R3884 EclX I from Enterobacter cloacae 590, Restriction Enzyme
R6265
EcoR I from Escherichia coli BS5, Restriction Enzyme
R2756
EcoR V from Escherichia coli, buffered aqueous glycerol solution
R5628 Hae III from Haemophilus aegyptius, Restriction Enzyme
R1632
Mva I from Micrococcus varians Rfl 19, Restriction Enzyme
R8761 Nco I from Nocardia corallina, Restriction Enzyme
R5634
Nhe I from Neisseria mucosa heidelbergensis, Restriction Enzyme
R8506
Not I from Nocardia otidiscaviarum, Restriction Enzyme
R5884 Nsi I from Neisseria sicca, Restriction Enzyme
R7023
Pst I from Providencia stuartii, Restriction Enzyme
R1508
Pvu I from Proteus vulgaris, Restriction Enzyme
R2631
Pvu II from Proteus vulgaris, Restriction Enzyme
R4756 Rsa I from Rhodopseudomonas sphaeroides, Restriction Enzyme
R0754 Sal I from Streptomyces albus G, Restriction Enzyme
R8256
Sfi I from Streptomyces fimbriatus, Restriction Enzyme
R7135 Sph I from Streptomyces phaeochromogenes, Restriction Enzyme
R7260
Xba I from Xanthomonas badrii, Restriction Enzyme
R6379
Xho I from Xanthomonas holcicola, Restriction Enzyme
R1259 Afl III from Anabaena flos-aquae, buffered aqueous glycerol solution