EMAIL THIS PAGE TO A FRIEND
Annales de dermatologie et de venereologie
[Efficacy of calcipotriol ointment in nevoid keratosis of the nipple].
PMID 23199772
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4369
Calcipotriol hydrate, ≥98% (HPLC)
C27H40O3 · xH2O