EMAIL THIS PAGE TO A FRIEND
Biophysical journal
Taking the pulse of protein interactions by EPR spectroscopy.
PMID 23200037
Abstract
An article by Gaffney et al. in this issue establishes a method using pulse electron paramagnetic resonance spectroscopy to determine the location of protein substrates or binding partners with high precision.