EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Topical application of Pleurotus eryngii extracts inhibits 2,4-dinitrochlorobenzene-induced atopic dermatitis in NC/Nga mice by the regulation of Th1/Th2 balance.
PMID 23200891
Abstract
Pleurotus eryngii is a nutritional and medicinal food rich in polysaccharides that enhance the host immune system as a response to various diseases. The present study investigated the effects of P. eryngii extracts (PEE) on the progress of atopic dermatitis (AD)-like skin lesions in NC/Nga mice induced by 2,4-dinitrochlorobenzene (DNCB). We evaluated skin dermatitis severity, ear thickness, histopathological examination, and cytokines level in DNCB-applied mice treated with PEE. Continuous treatment of PEE inhibited the development of the AD-like skin lesions. PEE suppressed DNCB-induced dermatitis severity, serum level of IgE and thymus and activation-regulated chemokine (TARC), and mRNA expression of TNF-α, INF-γ, IL-4, IL-5, and IL-13 in mice. In addition, PEE reduced thickness of the dermis and dermal infiltration of inflammatory cells and mast cells in histopathological examination. These results indicate that PEE inhibits allergic contact dermatitis through the modulating of T helper (Th)1 and Th2 responses and diminishing the inflammatory cells and mast cells infiltration in the skin lesions in NC/Nga mice.