EMAIL THIS PAGE TO A FRIEND
The Journal of hand surgery
Dupuytren contracture recurrence following treatment with collagenase clostridium histolyticum (CORDLESS study): 3-year data.
PMID 23200951
Abstract
To evaluate long-term efficacy and safety of collagenase clostridium histolyticum (CCH) after the third year of a 5-year nontreatment follow-up study, Collagenase Option for Reduction of Dupuytren Long-Term Evaluation of Safety Study. This study enrolled Dupuytren contracture patients from 5 previous clinical studies. Beginning 2 years after their first CCH injection, we re-evaluated patients annually for joint contracture and safety. Recurrence in a previously successfully treated joint (success = 0° to 5° contracture after CCH administration) was defined as 20° or greater worsening in contracture in the presence of a palpable cord or medical/surgical intervention to correct new or worsening contracture. We assessed partially corrected joints (joints reduced 20° or more from baseline contracture but not to 0° to 5°) for nondurable response, also defined as 20° or greater worsening of contracture or medical/surgical intervention. Of 1,080 CCH-treated joints (648 metacarpophalangeal [MCP]; 432 proximal interphalangeal [PIP]; n = 643 patients), 623 (451 MCP, 172 PIP) had achieved 0° to 5° contracture in the original study. Of these joints, 35% (217 of 623) recurred (MCP 27%; PIP 56%). Of these recurrences, an intervention was performed in 7%. Of the 1,080 CCH-treated joints, 301 were partially corrected in the original study. Of these, 50% (150 of 301; MCP: 38% [57 of 152]; PIP: 62% [93 of 149]) had nondurable response. We identified no new long-term or serious adverse events attributed to CCH during follow-up. Anti-clostridial type I collagenase and/or anti-clostridial type II collagenase antibodies were reported for 96% or more of patients who received 2 or more CCH injections and 82% who received 1 injection. The recurrence rate, which is comparable to other standard treatments, and the absence of long-term adverse events 3 years after initial treatment indicate that CCH is an effective and safe treatment for Dupuytren contracture. Most successfully treated joints had a contracture well below the threshold for surgical intervention 3 years after treatment. Recurrence rates among successfully treated joints were lower than nondurable response rates among partially corrected joints. Therapeutic IV.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1639
Collagenase from Clostridium histolyticum, sterile-filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C9891
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
C5894
Collagenase from Clostridium histolyticum, sterile; sterile-filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C2674
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
C9722
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), 0.5-5.0 FALGPA units/mg solid, cell culture tested
C1764
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat epididymal adipocytes tested, Type II-S, 0.5-5.0 FALGPA units/mg solid
C1889
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat hepatocytes tested, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C2139
Collagenase from Clostridium histolyticum, release of rat epididymal adipocytes and hepatocytes tested (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C9263
Collagenase from Clostridium histolyticum, release of physiologically active rat pancreatic islets tested, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
C2014
Collagenase from Clostridium histolyticum, sterile-filtered, suitable for release of physiologically active rat pancreatic islets, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
C7657
Collagenase from Clostridium histolyticum, release of physiologically active rat pancreatic islets tested, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
C9407
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
C9697
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), Suitable for digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
C0255
Collagenase from Clostridium histolyticum, purified by chromatography, ≥1 FALGPA units/mg solid, ≥250 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
C0773
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, 1,000-3,000 CDU/mg solid (CDU = collagen digestion units)
C2399
Collagenase from Clostridium histolyticum, sterile-filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, 1,000-3,000 CDU/mg solid
C2799
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, 1000-3000 CDU/mg solid (CDU = collagen digestion units), ≥4 FALGPA units/mg solid, high purity
C8051
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
C8176
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
C7926
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
C0374
Collagenase from Clostridium histolyticum, Animal component-free, ≥300 CDU/mg solid