EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Molybdenum enzymes, their maturation and molybdenum cofactor biosynthesis in Escherichia coli.
PMID 23201473
Abstract
Molybdenum cofactor (Moco) biosynthesis is an ancient, ubiquitous, and highly conserved pathway leading to the biochemical activation of molybdenum. Moco is the essential component of a group of redox enzymes, which are diverse in terms of their phylogenetic distribution and their architectures, both at the overall level and in their catalytic geometry. A wide variety of transformations are catalyzed by these enzymes at carbon, sulfur and nitrogen atoms, which include the transfer of an oxo group or two electrons to or from the substrate. More than 50 molybdoenzymes were identified in bacteria to date. In molybdoenzymes Mo is coordinated to a dithiolene group on the 6-alkyl side chain of a pterin called molybdopterin (MPT). The biosynthesis of Moco can be divided into four general steps in bacteria: 1) formation of the cyclic pyranopterin monophosphate, 2) formation of MPT, 3) insertion of molybdenum into molybdopterin to form Moco, and 4) additional modification of Moco with the attachment of GMP or CMP to the phosphate group of MPT, forming the dinucleotide variant of Moco. This review will focus on molybdoenzymes, the biosynthesis of Moco, and its incorporation into specific target proteins focusing on Escherichia coli. This article is part of a Special Issue entitled: Metals in Bioenergetics and Biomimetics Systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
203823
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Mo
510092
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Mo
266922
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Mo
266892
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Mo
357227
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Mo
577987
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Mo
266914
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis
Mo
366986
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Mo
357200
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Mo
GF93556693
Molybdenum, foil, 6mm disks, thickness 0.005mm, 99.9%
Mo
GF04293446
Molybdenum, tube, 500mm, outside diameter 3.15mm, inside diameter 2.39mm, wall thickness 0.38mm, 99.9%
Mo
GF30193781
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Mo
514802
Molybdenum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Mo
GF19350996
Molybdenum, wire reel, 5m, diameter 0.8mm, annealed, 99.95%
Mo
GF76876464
Molybdenum, wire reel, 1m, diameter 0.5mm, annealed, 99.95%
Mo
GF13141572
Molybdenum, foil, 6mm disks, thickness 0.5mm, annealed, 99.9%
Mo
GF68352330
Molybdenum, foil, 6mm disks, thickness 0.75mm, annealed, 99.9%
Mo
GF94586372
Molybdenum, foil, 8mm disks, thickness 0.005mm, 99.9%
Mo
GF30128576
Molybdenum, foil, 8mm disks, thickness 0.006mm, 99.9%
Mo
GF17537812
Molybdenum, foil, 8mm disks, thickness 0.007mm, 99.9%
Mo
GF40281803
Molybdenum, foil, 8mm disks, thickness 0.008mm, 99.9%
Mo
GF38800582
Molybdenum, foil, 8mm disks, thickness 0.009mm, 99.9%
Mo
GF04498702
Molybdenum, foil, 8mm disks, thickness 0.0125mm, 99.9%
Mo
GF88422127
Molybdenum, foil, 8mm disks, thickness 0.015mm, annealed, 99.9%
Mo
GF76305270
Molybdenum, foil, 8mm disks, thickness 0.015mm, as rolled, 99.9%
Mo
GF36011977
Molybdenum, foil, 8mm disks, thickness 0.01mm, 99.9%
Mo
GF93583402
Molybdenum, foil, 8mm disks, thickness 0.01mm, annealed, 99.9%
Mo
GF70653999
Molybdenum, foil, 8mm disks, thickness 0.020mm, annealed, 99.9%
Mo
GF77151629
Molybdenum, foil, 8mm disks, thickness 0.025mm, annealed, 99.9%
Mo
GF46109099
Molybdenum, foil, 8mm disks, thickness 0.03mm, annealed, 99.9%
Mo