EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Diol-linked microporous networks of cubic siloxane cages.
PMID 23203900
Abstract
A new class of inorganic-organic hybrid porous materials has been synthesized by a reaction between octa(hydridosilsesquioxane) (H(8)Si(8)O(12)), which has a double-four-ring (D4R) structure, and various diols, such as 1,3-propanediol (PD), 1,4-cyclohexanediol (CHD), and 1,3-adamantanediol (AD). Solid-state (29) Si magic-angle-spinning NMR spectroscopic analysis confirmed that most of the corner Si-H groups reacted with diols to form Si-O-C bonds with retention of the D4R cage. Nitrogen adsorption-desorption studies showed that the products are microporous solids with high BET surface areas (up to ≈580 m(2) g(-1) for CHD- and AD-linked products). If n-alkanediols are used as linkers, the surface area becomes smaller as the number of carbon atoms is increased. The thermal and hydrolytic stability of the products strongly depend on the type of diol linkers. The highest stabilities are found for the AD-linked products, which are thermally stable up to around 400 °C and remain intact even after being soaked in water for 1 day. In contrast, the PD-linked product is easily hydrolyzed in water to give microporous silica. These results offer a new route toward a series of silica-based porous materials with unique structures and properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C101206
1,4-Cyclohexanediol, 99%
C6H12O2