EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Ramariolides A-D, antimycobacterial butenolides isolated from the mushroom Ramaria cystidiophora.
PMID 23205944
Abstract
Four butenolides, ramariolides A-D (1-4), have been isolated from the fruiting bodies of the coral mushroom Ramaria cystidiophora. Their structures were elucidated by analysis of 1D and 2D NMR data and a single-crystal X-ray diffraction analysis of 1, and their absolute configurations were established using Mosher's method. The major metabolite, ramariolide A (1), which contains an unusual spiro oxiranebutenolide moiety, showed in vitro antimicrobial activity against Mycobacterium smegmatis and Mycobacterium tuberculosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
283754
2(5H)-Furanone, 98%
C4H4O2