EMAIL THIS PAGE TO A FRIEND
Chest
Management of recurrent malignant pleural effusions: an ever-recurring issue?
PMID 23208361
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86257
Talc, tested according to Ph Eur
H2Mg3O12Si4