EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Enantiospecific total synthesis of (-)-bengamide E.
PMID 23208822
Abstract
Total synthesis of the polyhydroxy caprolactam amide natural product, bengamide E, is accomplished starting from tartaric acid. Key reactions in the synthesis include desymmetrization of the bis(dimethylamide) unit of tartaric acid, Zn(BH(4))(2)-mediated anti-selective reduction, and a Horner-Wadsworth-Emmons olefination.