EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Direct catalytic asymmetric addition of allylic cyanides to aldehydes for expeditious access to enantioenriched unsaturated δ-valerolactones.
PMID 23208841
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122793
Allyl cyanide, 98%
C4H5N
389579
δ-Valerolactone, technical grade
C5H8O2