EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Draft genome sequence of Streptomyces sp. strain SS, which produces a series of uridyl peptide antibiotic sansanmycins.
PMID 23209237
Abstract
Streptomyces sp. SS produces a series of uridyl peptide antibiotic sansanmycins. Here, we present a draft genome sequence of Streptomyces sp. SS containing the biosynthetic gene cluster for the antibiotics. The identification of the biosynthetic gene cluster of sansanmycins may provide further insight into biosynthetic mechanisms for uridyl peptide antibiotics.