EMAIL THIS PAGE TO A FRIEND
Journal of pediatric hematology/oncology
Chemotherapy for initial induction failures in childhood acute lymphoblastic leukemia: a Children's Oncology Group Study (POG 8764).
PMID 23211688
Abstract
Children with acute lymphocytic leukemia who fail to enter remission have a poor prognosis. In a previous study, 9 of 14 children with induction failure entered remission after teniposide (VM26) plus cytosine arabinoside (Ara-C). We attempted to confirm these results. Twenty children received teniposide (200 mg/m/day IV) for 3 days and cytosine arabinoside (100 mg/m/day continuous IV infusion) for 7 days. There were 3 complete and 3 partial responses. Two additional patients achieved a complete response after a second, shorter course of the same agents. Although VM26 plus Ara-C is an active combination for treatment of acute lymphocytic leukemia induction failure, it does not appear as effective as in the initial report. Better treatments for this problem are needed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0609 Teniposide, ≥97% (HPLC)
C32H32O13S