EMAIL THIS PAGE TO A FRIEND
Pediatric dentistry
Clinical complications in the revascularization of immature necrotic permanent teeth.
PMID 23211919
Abstract
The purpose of this case series was to report on the use of a technique of revascularization for necrotic immature permanent teeth, several problems encountered, and solutions to those problems. Eighteen pulp revascularizations were performed in 2009 using the original protocol of revascularization (adapted from the AAE/AAPD joint meeting in 2007 in Chicago). The protocol consisted of opening the canal and disinfecting it with sodium hypochlorite, sealing in a triple antibiotic paste for 2-6 weeks, re-opening, re-irrigating, creating a blood clot in the canal, and sealing with an MTA barrier over the clot. Three problems were encountered during the treatment: (1) bluish discoloration of the crown; (2) failure to produce bleeding; and (3) collapse of the mineral trioxide aggregate (MTA) material into the canal. Modifications to solve these problems included: changing one of the antibiotics, using a local anesthesia without epinephrine, and adding collagen matrix to the blood clot.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6895
Cefaclor
C15H14ClN3O4S