EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Comparative analysis of selected biomarkers and pesticide sensitivity in juveniles of Solea solea and Solea senegalensis.
PMID 23212268
Abstract
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison of activities on enzymes involved in xenobiotic metabolism was carried out. Juveniles of both species were sampled in winter 2011 from the Ebro Delta region, and activities of selected enzymes such as acetylcholinesterase (AChE), carboxylesterase (CbE), ethoxyresorufin O-deethylase (EROD) and glutathione S-transferase (GST) were determined in several tissues. Lipid peroxidation (LP) levels in plasma were measured as a sign of oxidative stress. In vitro exposures to selected pesticides were contrasted, analysing AChE and CbE activities in several tissue homogenates. Overall, enzymatic activities were higher in S. solea except for gill GST and CbE and kidney GST, while plasmatic LP levels were similar. In vitro contrasts revealed lower IC50 values for CbE activities in S. solea, suggesting a greater buffer capacity of this enzyme to potentially reduce pesticide toxicity over AChE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0287 Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
E0162 Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
C4749 Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
46051 Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
79302 Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥8.0 U/mg
96667 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
51963 Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥0.8 U/mg
46069 Esterase from horse liver, lyophilized, powder, brown, 0.5-1.0 U/mg
46058 Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
E0887 Esterase from rabbit liver, lyophilized powder, ≥75 units/mg protein
79208 Esterase from Rhizopus oryzae, powder, light beige, ≥20 U/g
46071 Esterase from Saccharomyces cerevisiae, lyophilized, powder, beige, ~2 U/g
43388 Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
93855 Esterase Isoenzyme 2 porcine liver, recombinant, recombinant, expressed in E. coli, ≥80 U/g
93712 Esterase Isoenzyme 3 porcine liver, recombinant, recombinant, expressed in E. coli, ≥0.8 U/mg
53564 Esterase Isoenzyme 5 porcine liver, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
80712 Esterase Methylobacterium populi, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
92385 Esterase Nocardia farcinica, recombinant, recombinant, expressed in E. coli, ≥0.4 U/mg
04492 Esterase Paenibacillus barcinonensis, recombinant, recombinant, expressed in E. coli, ≥10 U/mg
04533 Esterase Pelobacter propionicus, recombinant, recombinant, expressed in E. coli, ≥0.5 U/mg
75742 Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
80711 Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥0.3 U/mg
62350 Lipozyme®, immobilized from Mucor miehei, granular, brown, >30 U/g