EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Analysis of petroleum fraction from flower, stem and leaf of Aconitum taipeicum by GC-MS].
PMID 23213736
Abstract
To analyze the chemical constituents of petroleum fraction of Aconitum taipeicum. The methanol extract of Aconitum taipeicum were extracted by petroleum and then analyzed by GC-MS. The compounds were quantitatively determined by normalization method. Twenty eight compounds were separated and identified. Most of them were alkane, fat acids and their esters and alkenes. The Nonacosane covered 13.057% of the total peaks, while 19-methyl-18,21-Hexatriacontanediether 8.180%, Ethylen eglycol monooctadecy ether 7.851%, 3,7,11,15-Tetramethyl-2-hexadecen-1-ol 7.805%, Metahyl Palmitate 6.676% and so on. This is the first report of constituents from the flower, stem and leaf of Aconitum taipeicum. The results will provide foundation for further exploitation and use of Aconitum taipeicum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74156
Nonacosane, analytical standard
C29H60
284246
Nonacosane, 99%
C29H60