EMAIL THIS PAGE TO A FRIEND
Veterinary surgery : VS
A biomechanical study of laparoscopic 4S-modified Roeder and Weston knot strength in 3-0 polyglactin 910 and 3-0 polydioxanone.
PMID 23215860
Abstract
To (1) evaluate biomechanical strength of 4 different laparoscopic knots using 2 suture types, and (2) evaluate carotid artery ligature bursting pressure of 2 knots using a single suture type. Biomechanical materials testing. Suture material (3-0 polydioxanone, 3-0 polyglactin 910). Four knot types were tested: 4S-modified Roeder (4SMR) Weston plus 3 square throws (W3S); Weston plus 3 granny throws (W3G); and a 4 square throw knot as a control (control); 24 specimens of each knot type were tied with 3-0 polyglactin 910 and 24 of 3 knot types (4SMR, W3S, control) were tied with 3-0 polydioxanone. Suture loop constructs were tested to 3 mm displacement failure and ultimate failure. Carotid artery ligation bursting pressure was tested using 10 samples each of 4SMR and W3S knots with 3-0 polyglactin 910. The W3S, W3G, and controls were similar. The 4SMR was similar to W3S using 3-0 PDS but the 4SMR had lower load to failure and greater elongation than the Weston using 3-0 polyglactin 910. The 4SMR had a higher slippage rate with 3-0 polyglactin 910. All ligatures withstood supraphysiologic pressures. Surgeons using 3-0 polyglactin 910 should consider using the Weston knot with added throws during laparoscopic procedures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2191
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
P1941
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
P2066
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
430471
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
719897
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 7,000-17,000
719889
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
719870
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
739952
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
719900
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
739944
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
739960
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000
719862
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
719919
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
719927
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
739979
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
719846
Resomer® X 206 S, poly(dioxanone)