EMAIL THIS PAGE TO A FRIEND
Pharmacogenomics
Evaluation of the effect of genetic variations in GATA-4 on the phenprocoumon and acenocoumarol maintenance dose.
PMID 23215884
Abstract
To investigate whether the phenprocoumon and acenocoumarol maintenance doses are influenced by genetic variations in GATA-4, a transcription factor of CYP2C9. The influence of seven GATA-4 SNPs on the coumarin maintenance dose was investigated by performing an analysis of variance trend analysis, stratified for CYP2C9 genotypes. Results of the best-explaining SNP were validated in the Rotterdam Study cohort. The largest dose differences were found for rs3735814 in patients using acenocoumarol and having the common allele for CYP2C9. The mean dosages decreased from 2.92 mg/day for the patients having the GATA-4 common alleles to 2.65 mg/day for the patients carrying one GATA-4 variant allele and to 2.37 mg/day for patients carrying two GATA-4 variant alleles (p = 0.004). Results could not be replicated in the validation cohort. For phenprocoumon, no significant effects were observed. Genetic variation in GATA-4 does not seem relevant for clinical implementation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0074
Acenocoumarol, ≥98% (HPLC)
C19H15NO6