EMAIL THIS PAGE TO A FRIEND
Experimental and clinical transplantation : official journal of the Middle East Society for Organ Transplantation
Modified BEAM conditioning regimen in patients with Hodgkin and non-Hodgkin lymphomas.
PMID 23216569
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5918
Lomustine, ≥98%
C9H16ClN3O2