EMAIL THIS PAGE TO A FRIEND
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer
Effects of β-lapachone on the production of Th1 and Th2 cytokines in C57BL/6 mice.
PMID 23216634
Abstract
In this study, we investigated the effects of β-lapachone (β-lap) on the production of cytokines in C57BL/6 mice. The culture supernatants of splenocytes exposed to β-lap plus lipopolysaccharide or concanavalin A (Con A) were harvested to determine Th1 (tumor necrosis factor-α, interferon-γ, interleukin [IL]-1β, IL-2, IL-17A, and IL-12) and Th2 (IL-4, IL-5, IL-6, and IL-10) cytokines. IL-2, a Th1 cytokine, was expressed at higher levels in splenocytes treated with β-lap and with lipopolysaccharide plus β-lap. Expression of Th2 cytokines, including IL-4, IL-5, IL-6, and IL-10, was lower in the culture supernatants of mouse splenocytes exposed to Con A plus β-lap than in supernatants from cells that were exposed to Con A alone. These findings demonstrate that β-lap downregulates the immediate hypersensitivity reaction induced by Con A. Therefore, Th2 responses involve the downregulation of IL-4, IL-5, IL-6, and IL-10, preventing the occurrence of allergies and various manifestations of allergic inflammation. These results suggest that β-lap may have potential preventive or adjunct anti-inflammatory therapy applications because of its function in modulating the production of cytokines. However, further in vivo investigations of this process are necessary to elucidate the mechanisms involved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2037
β-Lapachone, ≥98% (TLC)
C15H14O3