EMAIL THIS PAGE TO A FRIEND
Molecular pain
Galanin-expression and galanin-dependent sensory neurons are not required for itch.
PMID 23216829
Abstract
Galanin is a key modulator of nociception, and it is also required for the developmental survival of a subset of C-fibre sensory neurons which are critical to the mediation of neuropathic and inflammatory pain. However, the potential modulatory roles played by galanin, or the galanin-dependent neurons, in pruritoceptive mechanisms underlying the sensation of itch have not been investigated. Here we report that mice carrying a loss-of-function mutation in the galanin gene (Gal-KO) show no differences in spontaneous behavioural itch responses compared to wild-type (WT) controls. Similarly, the responses to a range of pruritogens are not significantly different between the two genotypes. These results suggest that neither galanin expression, nor the galanin-dependent subpopulation of sensory neurons is required for itch-related behaviours.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5773
Galanin porcine, ≥97% (HPLC), powder
C146H213N43O40