EMAIL THIS PAGE TO A FRIEND
Dermatology online journal
Topical iodophor preparations: chemistry, microbiology, and clinical utility.
PMID 23217942
Abstract
Iodophor preparations are commonly used in all medical specialties for antisepsis of the skin prior to injections, invasive procedures, and surgery. Povidone-iodine has some very intriguing properties that make it extremely effective as a broad spectrum bacteriocidal agent with no known bacterial resistance, potentially lending itself to broader applications than its current uses. In this article the background, formulations, chemistry, and microbiology of iodine will be reviewed and recent clinical investigations of utility beyond skin antisepsis will be discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PVP1
Poly(vinylpyrrolidone)–Iodine complex