EMAIL THIS PAGE TO A FRIEND
Current biology : CB
Evolutionary genetics: no coming back from neverland.
PMID 23218006
Abstract
Host-plant specialization plays a key role in insect evolution, but little is known about its molecular basis. A new paper shows that a cactus-feeding fly became restricted to its host by changes in an enzyme that converts dietary sterols into essential hormones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3652
5α-Cholest-7-en-3β-ol
C27H46O