EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Anti-inflammatory effect of fucoidan extracted from Ecklonia cava in zebrafish model.
PMID 23218269
Abstract
Fucoidan extracted from Ecklonia cava had strong anti-inflammatory activities. However, the direct effects of fucoidan of E. cava on anti-inflammatory activities in vivo model remained to be determined. Therefore, the present study was designed to assess in vivo anti-inflammatory effect of fucoidan extracted from E. cava (ECF) using tail-cutting-induced and lipopolysaccharide (LPS)-stimulated zebrafish model. Treating zebrafish model with tail-cutting and LPS-treatment significantly increased the ROS and NO level. However, ECF inhibited this tail-cutting-induced and LPS-stimulated ROS and NO generation. These results show that ECF alleviated inflammation by inhibiting the ROS and NO generation induced by tail-cutting and LPS-treatment. In addition, ECF has a protective effect against the toxicity induced by LPS exposure in zebrafish embryos. This outcome could explain the potential anti-inflammatory activity of ECF, which might have a beneficial effect during the treatment of inflammatory diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5631 Fucoidan from Fucus vesiculosus, Crude