EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Study of "napier grass" delignification for production of cellulosic derivatives.
PMID 23218375
Abstract
Recently, much research on the evaluation of new cellulose sources has been developed. In this context, a promising source is "napier grass", which contains 30.40% lignin, 36.34% cellulose, and 34.12% hemicellulose. In this work, conditions for the delignification of "napier grass" in the laboratory were studied by using calcium oxide (CaO) and hydrogen peroxide (H(2)O(2)). The best pulping conditions were 9.00% CaO for a period of 2.73 h, which resulted in 74.99% delignification and 66.58% cellulose. The best conditions for the bleaching process were pH 12 and hydrogen peroxide at concentration of 4.2% for 6h, at a temperature of 40 °C, which gave 90.98% delignification and 99.21% cellulose. The analyses were performed by using weight percent.