EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and biological evaluation of scopoletin derivatives.
PMID 23218780
Abstract
A series of new scopoletin derivatives were designed and synthesized. Their anti-proliferative effect was initially evaluated against various human cancer cell lines. Among the tested compounds, A1, A2, and D6 showed significant anti-proliferative activities. Angiogenesis was detected by endothelial cell migration assay and tube formation study. The results showed that A1, A2, and D6 inhibited the vascular endothelial growth factor (VEGF)-stimulated proliferation, migration, and tube formation of human umbilical vein endothelial cells in vitro. Moreover, they inhibited the vessel growth in the chorioallantoic membrane in vivo. This inhibition was correlated with a significant decrease in the VEGF-triggered phosphorylated forms of ERK1/2 and Akt. In summary, these findings strongly suggested that these scopoletin derivatives might be structurally novel angiogenesis inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2500
Scopoletin, ≥99%
C10H8O4
38332
Scopoletin, analytical standard
C10H8O4