EMAIL THIS PAGE TO A FRIEND
Phytomedicine : international journal of phytotherapy and phytopharmacology
Anti-tumor and anti-metastatic actions of wogonin isolated from Scutellaria baicalensis roots through anti-lymphangiogenesis.
PMID 23219337
Abstract
Tumor growth and metastasis are associated with angiogenesis and lymphangiogenesis through the production of vascular endothelial growth factor (VEGF) or VEGF-C in tumors, and the phosphorylation of VEGF receptor (VEGFR)-2 or VEGFR-3 in vascular endothelial cells or lymphatic endothelial cells (LECs). Tumor-associated macrophages (TAMs) play an important role in tumor lymphangiogenesis, and consequently stimulate metastasis through the lymphatic system to lymph nodes. We examined the effects of wogonin isolated from Scutellaria baicalensis roots on tumor growth and metastasis using a highly metastatic model in osteosarcoma LM8-bearing mice. Wogonin (25 and 50 mg/kg, twice daily) reduced tumor growth and metastasis to the lung, liver and kidney, angiogenesis (CD31-positive cells), lymphangiogenesis (LYVE-1-positive cells), and TAM (F4/80-positive cell) numbers in the tumors of LM8-bearing mice. Wogonin (10-100 μM) also inhibited increases in IL-1β production and cyclooxygenase (COX)-2 expression induced by lipopolysaccharide in THP-1 macrophages. Wogonin had no effect on VEGF-C production in LM8 cells, or VEGFR-3 expression in human lymphatic endothelial cells (HLECs), however, it inhibited VEGF-C-induced VEGFR-3 phosphorylation in HLECs. The anti-tumor and anti-metastatic actions of wogonin may be associated with the inhibition of VEGF-C-induced lymphangiogenesis through a reduction in VEGF-C-induced VEGFR-3 phosphorylation by the inhibition of COX-2 expression and IL-1β production in TAMs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W0769
Wogonin hydrate, ≥98% (HPLC), solid
C16H12O5 · xH2O