EMAIL THIS PAGE TO A FRIEND
Phytomedicine : international journal of phytotherapy and phytopharmacology
Reversal of dexamethasone induced insulin resistance in 3T3L1 adipocytes by 3β-taraxerol of Mangifera indica.
PMID 23219340
Abstract
The present study investigates the efficacy of Mangifera indica ethyl acetate extract (MIEE) and its bioactive compound, 3β-taraxerol in the reversal of dexamethasone (DEX) induced insulin resistance in 3T3L1 adipocytes. MIEE and 3β-taraxerol were evaluated for their ability to restore impaired glucose uptake and, expression of molecular markers in the insulin signaling pathway induced by DEX in 3T3L1 adipocytes using 2-deoxy-D-[1-(3)H] glucose uptake assay and ELISA. An insulin resistant model has been developed using a glucocorticoid, DEX on 3T3L1 adipocytes. Insulin resistant condition was observed at 24h of DEX induction wherein a maximum degree of resistance of about 50% was measured based on inhibition of glucose uptake, which was confirmed using cytotoxicity analysis. The developed model of insulin resistance was studied in comparison to positive control rosiglitazone. DEX induced inhibition of glucose uptake and the expression of insulin signaling markers GLUT4 and PI3K were found to be restored by 3β-taraxerol and MIEE, thus delineating its mechanism of action in the reversal of insulin resistance. 3β-Taraxerol effectively restored DEX induced desensitization via restoration of PI3K and GLUT4 expression. To conclude, since 3β-taraxerol exhibits significant effect in reversing insulin resistance it can be further investigated as an insulin resistance reversal agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75934
3β-Taraxerol, analytical standard
C30H50O