EMAIL THIS PAGE TO A FRIEND
The American Journal of Pathology
Overexpression of Rap-1A indicates a poor prognosis for oral cavity squamous cell carcinoma and promotes tumor cell invasion via Aurora-A modulation.
PMID 23219753