EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Anti-inflammatory and neuroprotective effects of auraptene, a citrus coumarin, following cerebral global ischemia in mice.
PMID 23219792
Abstract
Cerebral ischemia causes delayed neuronal cell death in the hippocampus resulting in sequential cognitive impairments. Hyper-activated inflammation following ischemia is one of the etiologies for delayed neuronal cell death. In the present study, using a transient global ischemia mouse model, we showed that auraptene (AUR), a citrus coumarin, effectively inhibited microglia activation, cyclooxygenase-2 expression by astrocytes, and neuronal cell death in the hippocampus following ischemic insults. These results suggest that AUR acts as a neuroprotective agent in the ischemic brain, which may be mediated by suppression of the inflammatory response.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9861
Auraptene, ≥98% (HPLC)
C19H22O3