EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Formation of enzyme polymer engineered structure for laccase and cross-linked laccase aggregates stabilization.
PMID 23220110
Abstract
Laccase and laccase-based cross-linked enzyme aggregates (CLEAs) were stabilized through the formation of a surrounding polymeric network made of chitosan and 3-aminopropyltriethoxysilane. The thermoresistance of the resulting enzyme polymer engineered structures of laccase (EPES-lac) and CLEAs (EPES-CLEA) were more than 30 times higher than that of free laccase and CLEAs at pH 3 and 40 °C. The EPES showed higher residual activity than the unmodified biocatalysts against chaotropic salts (up to 10 times), EDTA (up to 5 times), methanol (up to 15 times) and acetone (up to 20 times). The Michaelis-Menten kinetic parameters revealed that the affinity for 2,2'-azino-bis-(3-ethylbenzothiazoline-6-sulphonic acid) has doubled for the EPES-lac and EPES CLEA compared to their unmodified forms. The EPES-lac structures acted optimally at pH 4 and their activity was nearly temperature-independent, while the laccase activity of EPES-CLEA was optimal at pH 4 and 60 °C. Globally, the EPES have shown significantly improved properties which make them attractive candidate for the development of laccase-based applications.