EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Lucilia sericata chymotrypsin disrupts protein adhesin-mediated staphylococcal biofilm formation.
PMID 23220967
Abstract
Staphylococcus aureus and Staphylococcus epidermidis biofilms cause chronic infections due to their ability to form biofilms. The excretions/secretions of Lucilia sericata larvae (maggots) have effective activity for debridement and disruption of bacterial biofilms. In this paper, we demonstrate how chymotrypsin derived from maggot excretions/secretions disrupts protein-dependent bacterial biofilm formation mechanisms.