EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Cloning, heterologous expression, and enzymatic characterization of cathepsin L from starfish (Asterina pectinifera).
PMID 23221686
Abstract
To determine the role of cathepsin L in Echinoderms, starfish (Asterina pectinifera) cathepsin L (ApCtL) was cloned. The results of RT-PCR analysis indicated that the expression of ApCtL in all of the tissues. The pro-mature enzyme of ApCtL, proApCtL, was expressed in Escherichia coli, and cathepsin activity was detected by cleaving of synthetic fluorogenic peptide substrates and gelatin zymography.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6854 Cathepsin L from human liver, ≥0.5 units/mg protein, solution