EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis and antiradical/antioxidant activities of caffeic acid phenethyl ester and its related propionic, acetic, and benzoic acid analogues.
PMID 23222926
Abstract
Caffeic acid phenethyl ester (CAPE) is a bioactive component isolated from propolis. A series of CAPE analogues was synthesized and their antiradical/antioxidant effects analyzed. The effect of the presence of the double bond and of the conjugated system on the antioxidant effect is evaluated with the analogues obtained from 3-(3,4-dihydroxyphenyl) propanoic acid. Those obtained from 2-(3,4-dihydroxyphenyl) acetic acid and 3,4-dihydroxybenzoic acid allow the evaluation of the effect of the presence of two carbons between the carbonyl and aromatic system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
290009
Potassium benzoate, ReagentPlus®, 99%
C7H5KO2