EMAIL THIS PAGE TO A FRIEND
Skin therapy letter
Rosacea: update on management and emerging therapies.
PMID 23223767
Abstract
Rosacea is a common chronic skin disorder that has significant impact on the self-esteem and quality of life of affected individuals. Currently understood as an inflammatory condition that occurs in the context of an altered innate immune response, the available topical and systemic therapies function as immunomodulators to restore cutaneous homeostasis. The goals of therapy include reduction of papules, pustules, erythema and physical discomfort with improvement in quality of life. Standard topical treatments include metronidazole and azelaic acid, although many other agents and regimens have been presented. Subantimicrobial/antiinflammatory dose oral doxycycline was US FDA approved in 2006 for the management of rosacea, but Health Canada clearance was only recently granted for this indication. Furthermore, renewed research interest has led to the development of other emerging therapies including topical ivermectin, brimonidine and oxymetazoline that hold promise for patients suffering from this condition.