EMAIL THIS PAGE TO A FRIEND
The Journal of neuropsychiatry and clinical neurosciences
Acute onset of parkinsonism with reversible course after H1N1 vaccination: insight from a young lady.
PMID 23224477
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1260
Amantadine hydrochloride
C10H17N · HCl