EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Site-selective deuterated-alkene synthesis with palladium on boron nitride.
PMID 23225027
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
255475
Boron nitride, powder, ~1 μm, 98%
BN