EMAIL THIS PAGE TO A FRIEND
Inflammation
Neurogenic airway inflammation induced by repeated intra-esophageal instillation of HCl in guinea pigs.
PMID 23225164
Abstract
This study was conducted to investigate if repeated intra-esophageal acid administrations may induce neurogenic inflammation in the airways and nodose ganglion in a guinea pig model. Guinea pigs were sedated and perfused with 0.1xa0N HCl in the distal esophagus via a nasoesophageal catheter for 14 consecutive days. Substance P (SP), neurokinin A (NKA), neurokinin B (NKB), and calcitonin gene-related peptide concentration were measured by ELISA or radioimmunoassay. Neuropeptide expression in the airways and nodose ganglion was detected by immunohistochemistry and assessed semi-quantitatively. Inflammation was found in the trachea and bronchi. There was a threefold increase in substance P concentration in the trachea, main bronchi, and lung homogenate and a twofold increase in NKA and NKB concentration in the main bronchi, lung homogenate, and bronchial alveolus lavage fluid, respectively. The SP and NKA expressions in the airways and nodose ganglion were also significantly increased. Chronic intra-esophageal acid instillation induces significant neurogenic inflammation in the airways and nodose ganglion in the vagus nerve in guinea pigs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4267
Neurokinin A, ≥97% (HPLC)
C50H80N14O14S
N4143
Neurokinin B, ≥95% (HPLC)
C55H79N13O14S2