EMAIL THIS PAGE TO A FRIEND
Anticancer research
Effect of menatetrenone, a vitamin k2 analog, on recurrence of hepatocellular carcinoma after surgical resection: a prospective randomized controlled trial.
PMID 23225445
Abstract
The aim of this study was to investigate whether menatetrenone (MNT) suppresses hepatocellular carcinoma (HCC) recurrence in patients undergoing hepatectomy. Between January 2005 and September 2009, 101 patients who underwent curative hepatectomy for primary HCC were enrolled in the study. Patients were divided into two groups: a non-MNT group (n=51), and an MNT group (n=50) that was administered 45 mg of MNT daily. During the observation period, recurrence was observed in 33 patients in the non-MNT group and in 28 patients of the MNT group (p=0.545). In patients with a preoperative Des-γ-carboxy-prothrombin (DCP) level lower than 40 AU/l (n=38), the cumulative disease-free survival rates at 12, 36, and 60 months in the non-MNT group, were 81.3%, 0.0%, and 0.0%, respectively, while those in the MNT group were 78.3%, 58.1%, and 31.0%, respectively (p=0.060). MNT has a moderately suppressive effect on HCC recurrence after hepatectomy, especially in patients with a normal preoperative DCP level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47774
Menaquinone (K2), analytical standard
C31H40O2
V9378
Vitamin K2
C31H40O2