EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Adventures in bridgehead substitution chemistry: synthesis of polycyclic polyprenylated acylphloroglucinols (PPAPs).
PMID 23229029
Abstract
The polycyclic polyprenylated acylphloroglucinol (PPAP) family of natural products includes important compounds with notable biological activities, such as garsubellin A, hyperforin and clusianone. The synthesis of these complex, bridged, highly oxidized and substituted systems presents a formidable challenge to synthetic chemists. This feature article describes how the use of unconventional bridgehead substitution chemistry has enabled the synthesis of these natural products and their analogues.