EMAIL THIS PAGE TO A FRIEND
Experimental brain research
Using vertebral movement and intact paraspinal muscles to determine the distribution of intrafusal fiber innervation of muscle spindle afferents in the anesthetized cat.
PMID 23229776
Abstract
Increasing our knowledge regarding intrafusal fiber distribution and physiology of paraspinal proprioceptors may provide key insights regarding proprioceptive deficits in trunk control associated with low back pain and lead to more effective clinical intervention. The use of vertebral movement as a means to reliably stretch paraspinal muscles would greatly facilitate physiological study of paraspinal muscle proprioceptors where muscle tendon isolation is either very difficult or impossible. The effects of succinylcholine (SCh) on 194 muscle spindle afferents from lumbar longissimus or multifidus muscles in response to computer-controlled, ramp-and-hold movements of the L(6) vertebra were investigated in anesthetized cats. Paraspinal muscles were stretched by moving the L(6) vertebra 1.5-1.7 mm in the dorsal-ventral direction. Initial frequency (IF), dynamic difference (DD), their changes (∆) following SCh injection (100-400 μg kg(-1)), and post-SCh dynamic difference (SChDD) were measured. Muscle spindle intrafusal fiber terminations were classified as primary or secondary fibers as well as bag(1) (b(1)c), bag(2) (b(2)c), b(1)b(2)c, or chain (c) fibers. Intrafusal fiber subpopulations were distinguished using logarithmic transformation of SChDD and ∆IF distributions as established by previous investigators. Increases in DD indicate strength of b(1)c influence while increases in IF indicate strength of b(2)c influence. Out of 194 afferents, 46.9 % of afferents terminated on b(2)c fibers, 46.4 % on b(1)b(2)c fibers, 1 % on b(1)c fibers, and 5.7 % terminated on c fibers. Based on these intrafusal fiber subpopulation distributions, controlled vertebral movement can effectively substitute for direct tendon stretch and allow further investigation of paraspinal proprioceptors in this anatomically complex body region.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8251
Succinylcholine chloride dihydrate, 98.0-102.0%, solid
C14H30Cl2N2O4 · 2H2O