EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Application of diagnostic markers to invasive aspergillosis in children.
PMID 23231708
Abstract
Early mycological detection of Aspergillus species is the cornerstone for a prompt diagnosis, appropriate treatment strategies, and improved survival of patients with invasive aspergillosis (IA), irrespective of age. However, the currently available laboratory tests for the diagnosis of IA include culture with direct microscopy, histology, and antigenic markers, such as galactomannan and β-1, 3-d-glucan, Aspergillus spp. DNA detection by PCR, and imaging studies, such as high-resolution CT scan. However, all need further validation, especially in children. In this review we focus on the diagnosis of IA emphasizing the current perspectives, difficulties in interpretation, and the need of further evaluation from a pediatric point of view.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48230 D-Galacto-D-mannan from Ceratonia siliqua, ~95% (HPLC)