EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Yttrium phosphasalen initiators for rac-lactide polymerization: excellent rates and high iso-selectivities.
PMID 23231748
Abstract
Highly active yttrium phosphasalen initiators for the stereocontrolled ring-opening polymerization of rac-lactide are reported. The initiators are coordinated by a new class of ancillary ligand: an iminophosphorane derivative of the popular "salen" ligand, termed "phosphasalen". Changing the phosphasalen structure enables access to high iso-selectivities (P(i) = 0.84) or hetero-selectivities (P(s) = 0.87). The initiators also show very high rates, excellent polymerization control, and tolerance to low loadings; furthermore, no chiral auxiliaries/ligands are needed for the stereocontrol. The combination of such high rates with high iso-selectivities is very unusual.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
236071
N,N′-Bis(salicylidene)ethylenediamine, 98%
C16H16N2O2