EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Oxidative purification of halogenated ferrocenes.
PMID 23232493
Abstract
We report the large scale syntheses and 'oxidative purification' of fcI(2), fcBr(2) and FcBr (fc = ferrocene-1,1'-diyl, Fc = ferrocenyl). These valuable starting materials are typically laborious to separate via conventional techniques, but can be readily isolated by taking advantage of their increased E(1/2) relative to FcH/FcX contaminants. Our work extends this methodology towards a generic tool for the separation of redox active mixtures.