EMAIL THIS PAGE TO A FRIEND
Organic letters
An efficient, regioselective amination of 3,5-disubstituted pyridine N-oxides using saccharin as an ammonium surrogate.
PMID 23234245
Abstract
A process for the regioselective amination of unsymmetrical 3,5-substituted pyridine N-oxides has been developed utilizing cheap, readily available saccharin as an ammonium surrogate. High conversions of the corresponding saccharin adducts have been achieved under mild reaction conditions. In situ deprotection under acidic conditions allows for a one-pot process to substituted aminopyridines. High regioselectivities were obtained from a variety of 3,5-disubstituted pyridine N-oxides.