EMAIL THIS PAGE TO A FRIEND
Organic letters
Versatile routes to marine sponge metabolites through benzylidene rhodanines.
PMID 23234337
Abstract
The first total synthesis of the marine natural products Psammaplin C and Tokaradine A is described. Benzylidene rhodanines were utilized as versatile intermediates toward the synthesis of seven brominated marine sponge metabolites through the optimization of protection group strategies. Spermatinamine demonstrated good inhibition of all cancer cell lines tested, in particular the leukemia K562 and colon cancer HT29 cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83700
Rhodanine, for spectrophotometric det. of gallic acid, ≥99.0%
C3H3NOS2
118192
Rhodanine, 97%
C3H3NOS2