EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Synthesis of monoclinic potassium niobate nanowires that are stable at room temperature.
PMID 23234402
Abstract
We report the synthesis of KNbO(3) nanowires (NWs) with a monoclinic phase, a phase not observed in bulk KNbO(3) materials. The monoclinic NWs can be synthesized via a hydrothermal method using metallic Nb as a precursor. The NWs are metastable, and thermal treatment at ∼450 °C changed the monoclinic phase into the orthorhombic phase, which is the most stable phase of KNbO(3) at room temperature. Furthermore, we fabricated energy-harvesting nanogenerators by vertically aligning the NWs on SrTiO(3) substrates. The monoclinic NWs showed significantly better energy conversion characteristics than orthorhombic NWs. Moreover, the frequency-doubling efficiency of the monoclinic NWs was ∼3 times higher than that of orthorhombic NWs. This work may contribute to the synthesis of materials with new crystalline structures and hence improve the properties of the materials for various applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
541206
Potassium niobate
KNbO3